Learn More
Cayman Chemical Caspase-3humn Blockng Pe 200ug
SDP
Catalog No. 502911992
Encompass_Preferred
A typical antipsychotic binds to 5-HT6 and 5-HT7 receptors and the androgen receptor (Kis 72 2 4 and 800 nM respectively) inhibits the binding of SKF 10047 (Ki 36 nM) and spiperone (IC50 17 nM) to guinea pig brain membranes increases the duration of fighting and the latency to submission in mice
Catalog No. 50-291-1992 Supplier Cayman Chemical Supplier No. 360745200 UG
A typical antipsychotic binds to 5-HT6 and 5-HT7 receptors and the androgen receptor (Kis 72 2 4 and 800 nM respectively) inhibits the binding of SKF 10047 (Ki 36 nM) and spiperone (IC50 17 nM) to guinea pig brain membranes increases the duration of fighting and the latency to submission in mice