Learn More
Enzo Life Sciences Caspase-4 (human), (recombinant) (5000 U)
SDP
Supplier:  Enzo Life Sciences BMLSE1765000
Encompass_Preferred
Alternative name: ICH-2, Protease TX, ICErel-II. Concentration: 100 U/µl. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 50mM HEPES, pH 7.4, 100mM NaCl, 0.5% CHAPS, 1mM EDTA, 10% glycerol, 10mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-1951