Learn More
Enzo Life Sciences Caspase 4 monoclonal antibody (4B9) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM114E
Encompass_Preferred
Clone: 4B9. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human caspase-4 (N-terminal fragment). UniProt: P49662. Species Reactivity: Human. Applications: WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-658-711