Learn More
Apexbio Technology LLC Caspase-5-Fluoro-Assay-Kit
SDP
Catalog No. 5000567781
Encompass_Preferred
Caspase-5 Fluorometric Assay Kit is used in apoptosis and inflammation research to detect and measure caspase-5 activity which is part of the inflammatory response pathway Caspase-5 an enzyme in the caspase family plays a role in innate immune responses often studied in cell models like macrophages and epithelial cells The assay detects enzyme activity through fluorescence providing insights into pro-inflammatory cytokine production Useful for drug screening and disease models typical experimental concentrations depend on specific research designs In vitro activity is often observed in the nanomolar to low micromolar range
Catalog No. 50-005-67781 Supplier Apexbio Technology LLC Supplier No. K219525
Add to Cart
Add to Cart
Caspase-5 Fluorometric Assay Kit is used in apoptosis and inflammation research to detect and measure caspase-5 activity which is part of the inflammatory response pathway Caspase-5 an enzyme in the caspase family plays a role in innate immune responses often studied in cell models like macrophages and epithelial cells The assay detects enzyme activity through fluorescence providing insights into pro-inflammatory cytokine production Useful for drug screening and disease models typical experimental concentrations depend on specific research designs In vitro activity is often observed in the nanomolar to low micromolar range