Learn More
Enzo Life Sciences Caspase-5 (human), (recombinant) (5000 U)
SDP
Catalog No. 502011947
Encompass_Preferred
Alternative name: ICH-3, Protease TY, ICErel-III. Concentration: 100 U/µl. Purity: ≥80% (SDS-PAGE). Formulation: Liquid. In 50mM HEPES, pH 7.4, 100mM NaCl, 0.5% CHAPS, 1mM EDTA, 10% glycerol, 10mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-1947 Supplier Enzo Life Sciences Supplier No. BMLSE1715000
Alternative name: ICH-3, Protease TY, ICErel-III. Concentration: 100 U/µl. Purity: ≥80% (SDS-PAGE). Formulation: Liquid. In 50mM HEPES, pH 7.4, 100mM NaCl, 0.5% CHAPS, 1mM EDTA, 10% glycerol, 10mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.