Learn More
Enzo Life Sciences Caspase-7 monoclonal antibody (10-1-62) (25µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM137C
Encompass_Preferred
Clone: 10-1-62. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human Caspase-7 protein (full length). Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Monkey, Hamster. Applications: IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 10mM sodium phosphate, 50mM sodium chloride, and 0.1mM PMSF. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7137