Learn More
Enzo Life Sciences Caspase-7 monoclonal antibody (7-1-11) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM127E
Encompass_Preferred
Clone: 7-1-11. Host: Mouse. Isotype: IgG2b. Immunogen: Recombinant human Caspase-7 protein (full length). UniProt ID: P55210. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Dog. Specificity: Detects the unprocessed pro-caspase-7 (p35), caspase lacking the short amino-terminal pro-domain (p32) and fully processed caspase-7 (p19) in the apototic cell extract. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (0.2µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.1mM PMSF. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7136