Learn More
CPC Scientific MCA-Val-Asp-Gln-Val-Asp-Gly-Trp-Lys(DNP)-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108972
Encompass
Fluorogenic resonance energy transfer substrate used for the detection of caspase-7 activity.SEQUENCE: MCA-Val-Asp-Gln-Val-Asp-Gly-Trp-Lys(DNP)-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Mca-VDQVDGW-Lys(Dnp)-NH2MOLECULAR FORMULA: C60H74N14O21MOLECULAR WEIGHT: 1327.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:SKU(s): CASP-028A, CASP-028B
Catalog No. 50-210-8972 Supplier CPC Scientific Supplier No. CASP028A
Add to Cart
Add to Cart
Fluorogenic resonance energy transfer substrate used for the detection of caspase-7 activity.SEQUENCE: MCA-Val-Asp-Gln-Val-Asp-Gly-Trp-Lys(DNP)-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Mca-VDQVDGW-Lys(Dnp)-NH2MOLECULAR FORMULA: C60H74N14O21MOLECULAR WEIGHT: 1327.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:SKU(s): CASP-028A, CASP-028B