Learn More
Bioss Caspase-8 - C-terminal region
SDP
Catalog No. 5000065180
Encompass_Preferred
Caspase-8 (C-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB M382Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65180 Supplier Bioss Supplier No. BSM70376M
Caspase-8 (C-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB M382Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol