Learn More
Apexbio Technology LLC Caspase-8-Fluoro-Assay-Kit
SDP
Catalog No. 5000567605
Encompass_Preferred
Caspase-8 Fluorometric Assay Kit is a vital tool in apoptosis research originating from investigations into programmed cell death mechanisms It provides a clear pathway for detecting caspase-8 activity a key enzyme in the apoptosis signaling cascade especially the extrinsic pathway The kit measures enzyme activity using a fluorogenic substrate relevant for identifying apoptosis in various samples Common in vitro activity is often observed in the nanomolar to low micromolar range Typical applications include cell-based models animal studies and drug screening efforts facilitating the analysis of apoptotic processes This kit enables researchers to evaluate potential therapeutic agents and gain insights into disease mechanisms Experimental concentrations typically depend on the specific research design allowing customization for diverse experimental setups
Catalog No. 50-005-67605 Supplier Apexbio Technology LLC Supplier No. K2012200
Add to Cart
Add to Cart
Caspase-8 Fluorometric Assay Kit is a vital tool in apoptosis research originating from investigations into programmed cell death mechanisms It provides a clear pathway for detecting caspase-8 activity a key enzyme in the apoptosis signaling cascade especially the extrinsic pathway The kit measures enzyme activity using a fluorogenic substrate relevant for identifying apoptosis in various samples Common in vitro activity is often observed in the nanomolar to low micromolar range Typical applications include cell-based models animal studies and drug screening efforts facilitating the analysis of apoptotic processes This kit enables researchers to evaluate potential therapeutic agents and gain insights into disease mechanisms Experimental concentrations typically depend on the specific research design allowing customization for diverse experimental setups