Learn More
Enzo Life Sciences Caspase-8 (human) monoclonal antibody (12F5) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804242C100
Encompass_Preferred
Clone: 12F5. Isotype: IgG2b. Immunogen: Recombinant human caspase-8. Species reactivity: Human. Specificity: Recognizes procaspase-8 and active caspase-8. Detects procaspase-8 (p55/54), the intermediate cleavage products (p43/41) and the p18 active subunit of caspase-8 by Western blot. Applications: IHC (FS), IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In 0.15M PBS, pH 7.2, containing 1% BSA and 0.01% thimerosal.
Catalog No. 50-200-9839