Learn More
CPC Scientific Ac-Ile-Glu-Pro-Asp-CHO 1MG
SDP
Catalog No. 502109062
Encompass
ONE-LETTER SEQUENCE: Ac-IEPD-CHOMOLECULAR FORMULA: C22H34N4O9MOLECULAR WEIGHT:498.53STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: J.L. Harris et al., J. Biol. Chem., 273, 27364 (1998)
Catalog No. 50-210-9062 Supplier CPC Scientific Supplier No. CASP077A
ONE-LETTER SEQUENCE: Ac-IEPD-CHOMOLECULAR FORMULA: C22H34N4O9MOLECULAR WEIGHT:498.53STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: J.L. Harris et al., J. Biol. Chem., 273, 27364 (1998)