Learn More
CPC Scientific Ac-Ile-Glu-Pro-Asp-CHO 5MG
SDP
Catalog No. 502109063
Encompass
ONE-LETTER SEQUENCE: Ac-IEPD-CHOMOLECULAR FORMULA: C22H34N4O9MOLECULAR WEIGHT:498.53STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: J.L. Harris et al., J. Biol. Chem., 273, 27364 (1998)
Catalog No. 50-210-9063 Supplier CPC Scientific Supplier No. CASP077B
ONE-LETTER SEQUENCE: Ac-IEPD-CHOMOLECULAR FORMULA: C22H34N4O9MOLECULAR WEIGHT:498.53STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: J.L. Harris et al., J. Biol. Chem., 273, 27364 (1998)