Learn More
CPC Scientific Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Ile-Glu-Thr-Asp-CHO 0.5MG
SDP
Catalog No. 502109058
Encompass
Ac-AAVALLPAVLLALLAP-IETD-CHO is a composite of two peptide sequences, reversible caspase-8 inhibitor (Ac-IETD-CHO, CASP-057) and hydrophobic CCP signal peptide for Kaposi fibroblast growth factor (K-FGF).ONE-LETTER SEQUENCE: Ac-AAVALLPAVLLALLAPIETD-CHOMOLECULAR FORMULA: C95H162N20O26MOLECULAR WEIGHT:2000.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [886462-83-5], SYNONYMS: Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Ile-Glu-Thr-Asp-aldehyde, acetyl-AAVALLPAVLLALLAP-IETD-CHO, p-IETD-CHO, Ac-AAVALLPAVLLALLAP-IETD-CHORESEARCH AREA: ApoptosisREFERENCES: Lin, Yao-Zhong, et al. Journal of Biological Chemistry 270.24 (1995): 14255-14258. (original); Liang, Ye, Chaohua Yan, and Nina F. Schor. Oncogene 20.45 (2001): 6570.; Wondrak, Georg T. Free Radical Biology and Medicine 43.2 (2007): 178-190.; Satou, Motoyasu, et al. Endocrinology 151.10 (2010): 4765-4775.; Qiao, Shuxi, et al. Biochemi
Catalog No. 50-210-9058 Supplier CPC Scientific Supplier No. CASP075A
Ac-AAVALLPAVLLALLAP-IETD-CHO is a composite of two peptide sequences, reversible caspase-8 inhibitor (Ac-IETD-CHO, CASP-057) and hydrophobic CCP signal peptide for Kaposi fibroblast growth factor (K-FGF).ONE-LETTER SEQUENCE: Ac-AAVALLPAVLLALLAPIETD-CHOMOLECULAR FORMULA: C95H162N20O26MOLECULAR WEIGHT:2000.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [886462-83-5], SYNONYMS: Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Ile-Glu-Thr-Asp-aldehyde, acetyl-AAVALLPAVLLALLAP-IETD-CHO, p-IETD-CHO, Ac-AAVALLPAVLLALLAP-IETD-CHORESEARCH AREA: ApoptosisREFERENCES: Lin, Yao-Zhong, et al. Journal of Biological Chemistry 270.24 (1995): 14255-14258. (original); Liang, Ye, Chaohua Yan, and Nina F. Schor. Oncogene 20.45 (2001): 6570.; Wondrak, Georg T. Free Radical Biology and Medicine 43.2 (2007): 178-190.; Satou, Motoyasu, et al. Endocrinology 151.10 (2010): 4765-4775.; Qiao, Shuxi, et al. Biochemi