Learn More
Enzo Life Sciences Caspase-8 (mouse) monoclonal antibody (3B10) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804448C100
Encompass_Preferred
Clone: 3B10. Isotype: IgG1. Immunogen: Recombinant mouse caspase-8 p18 subunit. Species reactivity: Mouse. Applications: ELISA, Flow Cytometry, ICC, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9911