Learn More
CPC Scientific Ac-Ile-Glu-Thr-Asp-pNA 0.5MG
SDP
Catalog No. 502109090
Encompass
SEQUENCE: Ac-Ile-Glu-Thr-Asp-pNAONE-LETTER SEQUENCE: Ac-IETD-pNAMOLECULAR FORMULA: C27H38N6O12MOLECULAR WEIGHT: 638.63STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [219138-21-3]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:Grutter MG (2000) Curr Opin Struct Biol 10, 649-55Gastman BR (2001) Head Neck 23, 409-25Grutter MG (2000) Curr Opin Struct Biol 10, 649-55Stennicke HR and Salvesen GS (1999)Cell Death Differ 6, 1054-9Stennicke HR and Salvesen GS (1998) Biochim Biophys Acta 1387, 17-31Thornberry NA and Lazebnik Y (1998) Science 281, 1312-6Talanian RV, et al. (1997) . J Biol Chem 272, 9677-82.Fassy F, et al. (1998) Eur J Biochem 253, 76-83Datta R, et al. (1996) Blood 88, 1936-43Koeplinger KA, et al. (2000) Protein Expr Purif 18, 378-87SKU(s): CASP-090A, CASP-090B
Catalog No. 50-210-9090 Supplier CPC Scientific Supplier No. CASP090A
Add to Cart
Add to Cart
SEQUENCE: Ac-Ile-Glu-Thr-Asp-pNAONE-LETTER SEQUENCE: Ac-IETD-pNAMOLECULAR FORMULA: C27H38N6O12MOLECULAR WEIGHT: 638.63STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [219138-21-3]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:Grutter MG (2000) Curr Opin Struct Biol 10, 649-55Gastman BR (2001) Head Neck 23, 409-25Grutter MG (2000) Curr Opin Struct Biol 10, 649-55Stennicke HR and Salvesen GS (1999)Cell Death Differ 6, 1054-9Stennicke HR and Salvesen GS (1998) Biochim Biophys Acta 1387, 17-31Thornberry NA and Lazebnik Y (1998) Science 281, 1312-6Talanian RV, et al. (1997) . J Biol Chem 272, 9677-82.Fassy F, et al. (1998) Eur J Biochem 253, 76-83Datta R, et al. (1996) Blood 88, 1936-43Koeplinger KA, et al. (2000) Protein Expr Purif 18, 378-87SKU(s): CASP-090A, CASP-090B