Learn More
Enzo Life Sciences Caspase-9 (human), (recombinant) (5000 U)
SDP
Supplier:  Enzo Life Sciences BMLSE1735000
Encompass_Preferred
Alternative name: ICE-LAP-6, Mch6. Concentration: 100 U/µl. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 50mM HEPES, pH 7.4, 100mM NaCl, 0.5% CHAPS, 1mM EDTA, 10% glycerol, 10mM DTT. Source: Produced in E. coli. C-terminal Human caspase-9 (aa 131-416). Long Term Storage: -80°C.
Catalog No. 50-201-1949