Learn More
Medchemexpress LLC Caspase-9 inhibitor III (Ac-LEHD-cmk) | 403848-57-7 | 99.0% | 587.02 | C24H35ClN6O9 | 5 MG
SDP
Catalog No. 5000221844
Encompass_Preferred
Caspase-9 Inhibitor III (Ac-LEHD-cmk) is a selective, irreversible peptide-based inhibitor of caspase-9 used in biochemical and cellular research to block intrinsic apoptosis signaling. Supplied as a white to off-white solid with high reported purity, it is commonly used in studies of caspase-9-dependent pathways and models such as ischemia-reperfusion-induced myocardial injury. For research use only.
  • Selective, irreversible inhibition of caspase-9
  • Peptide-based inhibitor suitable for biochemical and cell-based assays
  • High purity for reproducible experimental results
  • Available in small quantities for assay optimization
  • Stable when stored frozen, reducing degradation risk
Catalog No. 50-002-21844 Supplier Medchemexpress LLC Supplier No. HY1367445MG
Caspase-9 Inhibitor III (Ac-LEHD-cmk) is a selective, irreversible peptide-based inhibitor of caspase-9 used in biochemical and cellular research to block intrinsic apoptosis signaling. Supplied as a white to off-white solid with high reported purity, it is commonly used in studies of caspase-9-dependent pathways and models such as ischemia-reperfusion-induced myocardial injury. For research use only.
  • Selective, irreversible inhibition of caspase-9
  • Peptide-based inhibitor suitable for biochemical and cell-based assays
  • High purity for reproducible experimental results
  • Available in small quantities for assay optimization
  • Stable when stored frozen, reducing degradation risk