Learn More
Cayman Chemical Caspase-9 Inhibitor III (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502932612
Encompass_Preferred
A caspase-9 inhibitor with cardioprotective effects; reduces infarct size, LDH and CK release, and DNA smearing induced by ischemia and reperfusion in isolated rat hearts; inhibits MMP-2-induced TnI proteolysis at 30 μM
Catalog No. 50-293-2612 Supplier Cayman Chemical Supplier No. 243061 MG
A caspase-9 inhibitor with cardioprotective effects; reduces infarct size, LDH and CK release, and DNA smearing induced by ischemia and reperfusion in isolated rat hearts; inhibits MMP-2-induced TnI proteolysis at 30 μM