Learn More
Enzo Life Sciences Caspase-9 monoclonal antibody (10-1-87) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905686100
Encompass_Preferred
Clone: 10-1-87. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant full length human Caspase-9. UniProt ID: P55211. Source: Purified from mouse ascites. Species reactivity: Human. Applications: WB. Recommended Dilutions: Western Blot (1µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide and 50% glycerol. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-6994