Learn More
Enzo Life Sciences Caspase-9 monoclonal antibody (5B4) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM139E
Encompass_Preferred
Clone: 5B4. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human caspase-9 (N-terminal fragment). UniProt ID: P55211. Species reactivity: Human, Mouse, Rat Drosophila. Applications: WB. Recommended Dilutions: Western Blot (1µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7139