Learn More
Enzo Life Sciences Caspase-9 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210815C100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 287-306 of human caspase-9. Species reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Hamster, Dog, Bovine. Applications: WB. Purity: Epitope-affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 30% glycerol, 0.5% BSA, 5mM EDTA, and 0.03% proclin. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.
Catalog No. 50-200-8164