Learn More
Enzo Life Sciences Caspase-9 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP149E
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human procaspase-9 (N-terminal fragment). UniProt ID: P55211. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Monkey, Guinea pig, Dog, Chicken, Bovine. Applications: WB. Recommended Dilutions: Western Blot (1:500, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7152