Learn More
Echelon Biosciences Research Labs Caspase Active Site Peptide - 5 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 866125MG
Encompass_Preferred
Caspase Active Site Peptide - 5mg , Ac-Ile-Gln-Ala-Cys-Arg-Gly-OH Category/Application: Enzyme Substrates This item is non returnable
Catalog No. 50-190-8940