Learn More
Enzo Life Sciences Caspase-Cell Lysis Buffer (30ml)
SDP
Catalog No. 502011089
Encompass_Preferred
Formulation: Liquid. In 50 mM HEPES, pH 7.4, containing 0.1% CHAPS, 5 mM DTT, 0.1 mM EDTA. Long Term Storage: -20°C.
Catalog No. 50-201-1089 Supplier Enzo Life Sciences Supplier No. BMLKI1170030
Formulation: Liquid. In 50 mM HEPES, pH 7.4, containing 0.1% CHAPS, 5 mM DTT, 0.1 mM EDTA. Long Term Storage: -20°C.