Learn More
ECM Bioscience Caspase Family Antibody Sampler Kit, ECM Biosciences
Catalog No. 502068629
Encompass_Preferred
The Caspase family antibody sampler kit can be used to detect the full length and cleaved products of Caspase-3, Caspase-6, Caspase-7, and Caspase-8. The kit also includes secondary reagent for detection of these antibodies. Applications: WB, E; Species Reactivity: Hu
Catalog No. 50-206-8629 Supplier ECM Bioscience Supplier No. CK6360
The Caspase family antibody sampler kit can be used to detect the full length and cleaved products of Caspase-3, Caspase-6, Caspase-7, and Caspase-8. The kit also includes secondary reagent for detection of these antibodies. Applications: WB, E; Species Reactivity: Hu