Learn More
Echelon Biosciences Research Labs Caspase 1 Substrate (ICE) fluorogenic (formerly IBCE substrate) - 5 mg, CAS#: 219137-85-6
SDP
Supplier:  Echelon Biosciences Research Labs 860175MG
Encompass_Preferred
Caspase 1 Substrate (ICE) fluorogenic (formerly IBCE substrate) - 5mg , Ac-Tyr-Val-Ala-Asp-AFC Category/Application: Enzyme Substrates This item is non returnable
Catalog No. 50-190-8860