Learn More
CPC Scientific Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Val-Ala-Asp-CHO 1MG
SDP
Catalog No. 502108991
Encompass
ONE-LETTER SEQUENCE: Ac-AAVALLPAVLLALLAPVAD-CHOMOLECULAR FORMULA: C88H151N19O22MOLECULAR WEIGHT:1827.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: N.A. Thornberry, and Y. Lazebnik, Science 281, 1312 (1998); Y-Z Lin, et al., J. Biol. Chem., 271, 5305 (1996); Y-Z Lin, et al., J. Biol. Chem., 270, 14255 (1995)
Catalog No. 50-210-8991 Supplier CPC Scientific Supplier No. CASP042B
ONE-LETTER SEQUENCE: Ac-AAVALLPAVLLALLAPVAD-CHOMOLECULAR FORMULA: C88H151N19O22MOLECULAR WEIGHT:1827.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: N.A. Thornberry, and Y. Lazebnik, Science 281, 1312 (1998); Y-Z Lin, et al., J. Biol. Chem., 271, 5305 (1996); Y-Z Lin, et al., J. Biol. Chem., 270, 14255 (1995)