Learn More
Enzo Life Sciences Caspase-1 assay kit for drug discovery (1x96wells)
SDP
Supplier:  Enzo Life Sciences BMLAK7010001
Encompass_Preferred
NOW INCLUDES BOTH COLORIMETRIC AND FLUOROGENIC SUBSTRATES! This kit is a complete assay system to measure protease activity of recombinant caspase-1. Cleavage of a tetrapeptide substrate is monitored colorimetrically at 405 nm or fluorimetrically (Ex. 360 nm/Em. 460 nm). Assays are performed in a convenient 96-well plate format. Applications: Colorimetric detection, Fluorescent detection, HTS Activity assay.
Catalog No. 50-201-0296