Learn More
AdipoGen anti-Caspase-1 (p10) (mouse), mAb (Casper-2) (Biotin) (100 µg)
SDP
Supplier:  AdipoGen AG20B0044BC100
Encompass_Preferred
Monoclonal Antibody. Recognizes endogenous full-length and activated (p10 fragment) mouse caspase-1. Isotype: Mouse IgG2a. Clone: Casper-2. Applications: WB. Liquid. In PBS containing 0.02% sodium azide. Caspase-1 is the best-described inflammatory caspase. It processes the cytokines interleukin-1beta (IL-1beta) and IL-18 and induces pyroptotic cell death. Caspase-1 is activated by multiprotein complexes called inflammasomes in response to numerous stimuli that are detected through distinct inflammasomes. NLRC4 responds to cytosolic flagellin, murine NLRP1b responds to anthrax lethal toxin, AIM2 responds to cytosolic DNA and NLRP3 responds to a variety of agonists including crystals.
Catalog No. 50-205-2955