Learn More
Cayman Chemical Caspase-3/7 Cell-Based Activity Assay Kit, 96 Well
SDP
Catalog No. 502895281
Encompass_Preferred
Cayman’s Caspase-3/7 Cell-Based Activity Assay Kit provides a convenient method of detecting caspase-3 and caspase-7 activity in cell lysates. Activity is monitored using a caspase-3/7 specific substrate, N-Ac-DEVD-AFC. Cleavage by the active enzyme(s) generates a highly fluorescent product that can be measured using excitation and emission wavelengths of 400 and 505 nm, respectively.Active caspase-3 is included in the kit as a positive control and staurosporine is included to induce apoptosis. The caspase-3/7 inhibitor, N-Ac-DEVD-CHO, is included as a control to confirm the specificity of the fluorescence signal to the activity of the enzymes.
Catalog No. 50-289-5281 Supplier Cayman Chemical Supplier No. 70279096 WELL
Cayman’s Caspase-3/7 Cell-Based Activity Assay Kit provides a convenient method of detecting caspase-3 and caspase-7 activity in cell lysates. Activity is monitored using a caspase-3/7 specific substrate, N-Ac-DEVD-AFC. Cleavage by the active enzyme(s) generates a highly fluorescent product that can be measured using excitation and emission wavelengths of 400 and 505 nm, respectively.Active caspase-3 is included in the kit as a positive control and staurosporine is included to induce apoptosis. The caspase-3/7 inhibitor, N-Ac-DEVD-CHO, is included as a control to confirm the specificity of the fluorescence signal to the activity of the enzymes.