Learn More
AdipoGen anti-Caspase-4 /11 (p20), mAb (Flamy-1) (Biotin) (100 µg)
SDP
Supplier:  AdipoGen AG20B0060BC100
Encompass_Preferred
Monoclonal Antibody. Recognizes endogenous full-length protein and activated (p20) fragment of mouse and human caspase-4/11. Isotype: Mouse IgG2bkappa. Clone: Flamy-1. Applications: IP, WB. Liquid. In PBS containing 0.02% sodium azide. Unlike caspase-1, the expression of caspase-11 is LPS-inducible and it is reasonable to postulate that other members of the family are regulated at the transcriptional or translational level by extracellular stimuli. Recently caspase-11 has been shown to be required for activation of the Nlrp3 inflammasome upon E. coli, V. cholerae and C. rodentium infection. The upstream activators of caspase-11 are TLR4 and TRIF, that modulate enteropathogen-induced inflammasome activation by promoting caspase-11 expression and activation. Formation of ASC foci (specks), a measure of NLRP3/ASC complex formation, requires caspase-11 but not caspase-1 indicating that caspase-11 acts upstream of the NLRP3/ASC complex.
Catalog No. 50-205-2972