Learn More
Cayman Chemical Caspase6 InhIbItrAcVEID 25uL
SDP
Catalog No. 502896175
Encompass_Preferred
An ionizable cationic lipid LNPs containing IZ-chol can be complexed with DNA with the plasmid-binding efficacy dependent in part on the type of helper lipids in the LNP formulation
Catalog No. 50-289-6175 Supplier Cayman Chemical Supplier No. 70189325 UL
An ionizable cationic lipid LNPs containing IZ-chol can be complexed with DNA with the plasmid-binding efficacy dependent in part on the type of helper lipids in the LNP formulation