Learn More
Medchemexpress LLC HY-17006 10mg Medchemexpress, Caspofungin (Acetate) CAS:179463-17-3 Purity:>98%
SDP
Catalog No. 502029226
Encompass_Preferred
Medchemexpress, HY-17006 10mg Caspofungin (Acetate) CAS:179463-17-3 Caspofungin Acetate (MK-0991 Acetate) is an antifungal drug, and noncompetitively inhibits 1,3-β-d glucan synthase activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9226 Supplier Medchemexpress LLC Supplier No. HY1700610MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-17006 10mg Caspofungin (Acetate) CAS:179463-17-3 Caspofungin Acetate (MK-0991 Acetate) is an antifungal drug, and noncompetitively inhibits 1,3-β-d glucan synthase activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.