Learn More
Apexbio Technology LLC Caspofungin Acetate 179463-17-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062053
Encompass_Preferred
Caspofungin acetate (MK-0991) is an antifungal lipopeptide compound targeting fungal cell wall synthesis by inhibiting 1 3- -D-glucan synthase thereby disrupting structural integrity This inhibition mechanism leads to the blockage of glucan polymer production essential for fungal growth In vitro analyses indicate Caspofungin acetate demonstrates antifungal activity against species including Aspergillus fumigatus and several Candida species with MIC90 values around 0 5 g/mL for C albicans and C parapsilosis 1 0 g/mL for C glabrata and C tropicalis and 2 0 g/mL for C krusei Caspofungin is utilized in biomedical research to explore cell wall synthesis pathways fungal susceptibility tests and mechanistic antifungal studies
Catalog No. 50-000-62053 Supplier Apexbio Technology LLC Supplier No. B20835.1
Caspofungin acetate (MK-0991) is an antifungal lipopeptide compound targeting fungal cell wall synthesis by inhibiting 1 3- -D-glucan synthase thereby disrupting structural integrity This inhibition mechanism leads to the blockage of glucan polymer production essential for fungal growth In vitro analyses indicate Caspofungin acetate demonstrates antifungal activity against species including Aspergillus fumigatus and several Candida species with MIC90 values around 0 5 g/mL for C albicans and C parapsilosis 1 0 g/mL for C glabrata and C tropicalis and 2 0 g/mL for C krusei Caspofungin is utilized in biomedical research to explore cell wall synthesis pathways fungal susceptibility tests and mechanistic antifungal studies