Learn More
TARGETMOL CHEMICALS INC CASPOFUNGIN ACETATE 25MG
SDP
Catalog No. 502581091
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Caspofungin Acetate (MK 0991) is the acetate salt of an antimycotic echinocandin lipopeptide semisynthetically derived from a fermentation product of the fungus Glarea lozoyensis. This agent is active against Aspergillus and Candida species. purity: 99%
Catalog No. 50-258-1091 Supplier TARGETMOL CHEMICALS INC Supplier No. T179925MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Caspofungin Acetate (MK 0991) is the acetate salt of an antimycotic echinocandin lipopeptide semisynthetically derived from a fermentation product of the fungus Glarea lozoyensis. This agent is active against Aspergillus and Candida species. purity: 99%