Learn More
Medchemexpress LLC HY-17006 1g Medchemexpress, Caspofungin (Acetate) L 743872;L 743873;MK 0991 CAS:179463-17-3 Purity:98%
SDP
Catalog No. 501638591
Encompass_Preferred
Medchemexpress,HY-17006 1g Caspofungin (Acetate) L 743872;L 743873;MK 0991 CAS:179463-17-3 Formula:C56H96N10O19 Caspofungin (Acetate) is an antifungal drug, and noncompetitively inhibits 1,3-β-d glucan synthase activity. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8591 Supplier Medchemexpress LLC Supplier No. HY170061G
Medchemexpress,HY-17006 1g Caspofungin (Acetate) L 743872;L 743873;MK 0991 CAS:179463-17-3 Formula:C56H96N10O19 Caspofungin (Acetate) is an antifungal drug, and noncompetitively inhibits 1,3-β-d glucan synthase activity. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.