Learn More
Medchemexpress LLC Caspofungin-d4 | 1131958-73-0 | 1097.34 | 500 UG
SDP
Catalog No. 5000390713
Encompass_Preferred
Caspofungin-d4 is a deuterium-labeled version of Caspofungin, a potent antifungal agent. It works by inhibiting the synthesis of β-(l,3)-D-glucan, a crucial component of the fungal cell wall. This compound is intended for research use only and is not for patient administration. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled.
  • Potent antifungal agent.
  • Inhibits synthesis of fungal cell wall component β-(l,3)-D-glucan.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90713 Supplier Medchemexpress LLC Supplier No. HY17006AS500UG
Caspofungin-d4 is a deuterium-labeled version of Caspofungin, a potent antifungal agent. It works by inhibiting the synthesis of β-(l,3)-D-glucan, a crucial component of the fungal cell wall. This compound is intended for research use only and is not for patient administration. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled.
  • Potent antifungal agent.
  • Inhibits synthesis of fungal cell wall component β-(l,3)-D-glucan.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.