Learn More
Cayman Chemical CaspofungIn-d4acetate 500ug
SDP
Catalog No. 502938231
Encompass_Preferred
An internal standard for the quantification of caspofungin by GC- or LC-MS
Catalog No. 50-293-8231 Supplier Cayman Chemical Supplier No. 29096500 UG
An internal standard for the quantification of caspofungin by GC- or LC-MS