Learn More
Medchemexpress LLC Caspofungin diacetate | 179463-17-3 | 99.7% | 1213.42 | 100 MG
SDP
Catalog No. 5000391252
Encompass_Preferred
Caspofungin diacetate is a potent antifungal agent. It functions by inhibiting the synthesis of β-(l,3)-D-glucan, a key component of the fungal cell wall.
  • Potent antifungal agent.
  • Inhibits the synthesis of the fungal cell wall component β-(l,3)-D-glucan.
  • For research use only.
Catalog No. 50-003-91252 Supplier Medchemexpress LLC Supplier No. HY17006100MG
Caspofungin diacetate is a potent antifungal agent. It functions by inhibiting the synthesis of β-(l,3)-D-glucan, a key component of the fungal cell wall.
  • Potent antifungal agent.
  • Inhibits the synthesis of the fungal cell wall component β-(l,3)-D-glucan.
  • For research use only.