Learn More
Medchemexpress LLC Caspofungin diacetate | 179463-17-3 | 99.79% | 1213.42 | 200 MG
SDP
Catalog No. 5000391296
Encompass_Preferred
Caspofungin diacetate, also known as MK-0991 diacetate or L-743872 diacetate, is a potent antifungal agent. Its mechanism of action involves inhibiting the synthesis of β-(l,3)-D-glucan, a crucial component of the fungal cell wall. This product is strictly for research use.
  • Potent antifungal activity.
  • Inhibits fungal cell wall synthesis by targeting β-(l,3)-D-glucan.
  • Demonstrates central nervous system penetration in mice, leading to a reduction in Candida burden in the brain at dosages of 1-8 mg/kg (intraperitoneal, daily for 7 days).
  • Exhibits a favorable safety profile at vitreal concentrations of 0.41 to 4.1 μM in mice, with nonsignificant alterations in ERG waveforms.
  • Widely cited in numerous publications, affirming its utility in research applications.
Catalog No. 50-003-91296 Supplier Medchemexpress LLC Supplier No. HY17006200MG
Caspofungin diacetate, also known as MK-0991 diacetate or L-743872 diacetate, is a potent antifungal agent. Its mechanism of action involves inhibiting the synthesis of β-(l,3)-D-glucan, a crucial component of the fungal cell wall. This product is strictly for research use.
  • Potent antifungal activity.
  • Inhibits fungal cell wall synthesis by targeting β-(l,3)-D-glucan.
  • Demonstrates central nervous system penetration in mice, leading to a reduction in Candida burden in the brain at dosages of 1-8 mg/kg (intraperitoneal, daily for 7 days).
  • Exhibits a favorable safety profile at vitreal concentrations of 0.41 to 4.1 μM in mice, with nonsignificant alterations in ERG waveforms.
  • Widely cited in numerous publications, affirming its utility in research applications.