Learn More
Medchemexpress LLC Caspofungin (diacetate) | 179463-17-3 | 99.8% | 1213.42 | 25 MG
SDP
Catalog No. 5000393123
Encompass_Preferred
Caspofungin diacetate is a potent antifungal agent that inhibits the synthesis of the fungal cell wall component β-(1,3)-D-glucan. It is suitable for laboratory research applications.
  • Potent antifungal agent
  • Inhibits the synthesis of the fungal cell wall component β-(1,3)-D-glucan
  • Penetrates the CNS in mice and reduces Candida burden in the brain (in vivo study)
  • Safe antifungal agent at vitreal concentrations of 0.41 to 4.1 μM in mice (in vivo study)
Catalog No. 50-003-93123 Supplier Medchemexpress LLC Supplier No. 25MGHY17006
Caspofungin diacetate is a potent antifungal agent that inhibits the synthesis of the fungal cell wall component β-(1,3)-D-glucan. It is suitable for laboratory research applications.
  • Potent antifungal agent
  • Inhibits the synthesis of the fungal cell wall component β-(1,3)-D-glucan
  • Penetrates the CNS in mice and reduces Candida burden in the brain (in vivo study)
  • Safe antifungal agent at vitreal concentrations of 0.41 to 4.1 μM in mice (in vivo study)