Learn More
Sklar Instruments Instrument Cassette, Sklar, 7 x 5 x 6/7 in. Instrument Cassette Medium with Hinged Lid and Silicone Instrument Holders, Holds Max. 10 Instruments, Sklar's Instrument Cassettes are used for cleaning, storage and sterilization. EA/1.
SDP
Catalog No. 501437225 Shop All Sklar Instruments Products
Encompass_Preferred
Instrument Cassette, Sklar, 7 x 5 x 6/7 in. Instrument Cassette Medium with Hinged Lid and Silicone Instrument Holders, Holds Max. 10 Instruments, Sklar's Instrument Cassettes are used for cleaning, storage and sterilization. The silicone supports are repositionable to assist in arranging various instruments. The cassettes are available with a hinged or detachable lid. This product is a medium cassette with a hinged lid and can hold approximately 10 instruments.
This item is not returnable. View return policy
Catalog No. 50-143-7225 Supplier Sklar Instruments Supplier No. 106511
Instrument Cassette, Sklar, 7 x 5 x 6/7 in. Instrument Cassette Medium with Hinged Lid and Silicone Instrument Holders, Holds Max. 10 Instruments, Sklar's Instrument Cassettes are used for cleaning, storage and sterilization. The silicone supports are repositionable to assist in arranging various instruments. The cassettes are available with a hinged or detachable lid. This product is a medium cassette with a hinged lid and can hold approximately 10 instruments.