Learn More
Newcomer Supply Cassette Basket, Deluxe
SDP
Catalog No. NC1393260
Encompass
The Deluxe Cassette Baskets holds up to 150 cassettes stacked vertically (label side up). It has removable dividers and a hinged lid that locks into place. Dimensions are identical to Sakura basket: 9" x 6" x 2 1/8" (H)
Catalog No. NC1393260 Supplier Newcomer Supply Supplier No. 5112C
The Deluxe Cassette Baskets holds up to 150 cassettes stacked vertically (label side up). It has removable dividers and a hinged lid that locks into place. Dimensions are identical to Sakura basket: 9" x 6" x 2 1/8" (H)