Learn More
Newcomer Supply Cassette Basket, Medium
SDP
Catalog No. NC1244158
Encompass
The Medium Cassette Basket holds up to 100 cassettes stacked horizontally. It has removable dividers and a lid with handle (lid does not lock in place). Dimensions are identical to Sakura basket: 9" x 5 1/2" x 1 1/4" (H)
Catalog No. NC1244158 Supplier Newcomer Supply Supplier No. 5112B
The Medium Cassette Basket holds up to 100 cassettes stacked horizontally. It has removable dividers and a lid with handle (lid does not lock in place). Dimensions are identical to Sakura basket: 9" x 5 1/2" x 1 1/4" (H)