Learn More
Newcomer Supply Cassette Basket, Rapid Processing
SDP
Catalog No. NC1927436
Encompass
The Rapid Processing Cassette Basket holds up to 76 cassettes stacked vertically (label side up). The basket has a snap in place lid with handle. Dimensions are identical to the Leica spaced basket: 5 1/4" x 7 1/2" x 1 3/4" (H)
Catalog No. NC1927436 Supplier Newcomer Supply Supplier No. 5112D
The Rapid Processing Cassette Basket holds up to 76 cassettes stacked vertically (label side up). The basket has a snap in place lid with handle. Dimensions are identical to the Leica spaced basket: 5 1/4" x 7 1/2" x 1 3/4" (H)