Learn More
Electron Microscopy Sciences HistoCool™ Tissue Cassette Cooling Tray Large, Blue
SDP
Catalog No. 501928646
Encompass_Preferred
These trays effectively aid in the cooling of tissue blocks. They are filled with freezer gel that stays frozen longer, up to 6-8 hours. Simply refreeze at the end of each day.
Color: Blue
Capacity: 30 cassette
Dimensions: 12.5(L) x 8.25 (W) x 4 (H)
Catalog No. 50-192-8646 Supplier Electron Microscopy Sciences Supplier No. 633142
These trays effectively aid in the cooling of tissue blocks. They are filled with freezer gel that stays frozen longer, up to 6-8 hours. Simply refreeze at the end of each day.
Color: Blue
Capacity: 30 cassette
Dimensions: 12.5(L) x 8.25 (W) x 4 (H)