Learn More
Electron Microscopy Sciences HistoCool™ Tissue Cassette Cooling Tray Small, Red
SDP
Catalog No. 501928645
Encompass_Preferred
These trays effectively aid in the cooling of tissue blocks. They are filled with freezer gel that stays frozen longer, up to 6-8 hours. Simply refreeze at the end of each day.
Color: Red
Capacity: 20 cassette
Dimensions: 8.5(L) x 8 (W) x 3 (H)
Catalog No. 50-192-8645 Supplier Electron Microscopy Sciences Supplier No. 633141
These trays effectively aid in the cooling of tissue blocks. They are filled with freezer gel that stays frozen longer, up to 6-8 hours. Simply refreeze at the end of each day.
Color: Red
Capacity: 20 cassette
Dimensions: 8.5(L) x 8 (W) x 3 (H)