Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LP Italiana Spa™ Embedding Cassette with Lid
GSA_VA

Catalog No. 50868790 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Embedding Cassette
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
50-868-790 Embedding Cassette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-868-790 Supplier LP Italiana Spa™ Supplier No. 191022
Add to Cart
Add to Cart

White embedding cassette with lid for pathological embedding.

White embedding cassette with lid for pathological embedding.

Embedding cassette for use in the pathological embedding proccess.

  • With lid
  • No ring
  • Great for use with tissue samples

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Embedding Cassette
Color White
For Use With (Application) Embedding
Includes Lid
Quantity 250 Pk.
Weight (English) 0.13 oz.
Weight (Metric) 3.72 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.