Learn More
Perkin Elmer US LLC Cassette for Avio 200 Torch Body Assembly
Catalog No. 502112410
Encompass_Preferred
This cassette is included in the Avio 200 Adjustable Torch Body Assembly (N0790138). This assembly also includes a Torch Mount (N0791285).
Catalog No. 50-211-2410 Supplier Perkin Elmer US LLC Supplier No. N0790214
This cassette is included in the Avio 200 Adjustable Torch Body Assembly (N0790138). This assembly also includes a Torch Mount (N0791285).